accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 113170-71-1

NH2BrNOOFFF

CAS RN 113170-71-1

IUPAC Name: 2-bromo-6-nitro-4-(trifluoromethyl)aniline
Synonyms: 2-bromo-6-nitro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 285.02
Molecular Formula: C7H4BrF3N2O2
InChi Key: ZUZMWPRSGJTLHW-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(C=C1[N+]([O-])=O)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products