CAS RN 113193-60-5
CAS RN 113193-60-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Iron(II) ethylenediammonium sulfate tetrahydrate, 99%
CAS: 113193-60-5 MDL Number: MFCD00082474