accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 113421-98-0

NH2OFFFF

CAS RN 113421-98-0

IUPAC Name: 4-fluoro-3-(trifluoromethoxy)aniline
Synonyms: 4-fluoro-3-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 195.12
Molecular Formula: C7H5F4NO
InChi Key: JSMGZPOPIVVIRZ-UHFFFAOYSA-N
SMILES: NC1=CC(OC(F)(F)F)=C(F)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products