accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1135278-54-4

CH3HNHNOONHBrPOOHHOHCl

CAS RN 1135278-54-4

IUPAC Name: (1-{[(7-bromo-2,3-dioxo-1,2,3,4-tetrahydroquinoxalin-5-yl)methyl]amino}ethyl)phosphonic acid hydrochloride
Synonyms: 1-{[(7-bromo-2,3-dioxo-1,4-dihydroquinoxalin-5-yl)methyl]amino}ethylphosphonic acid hydrochloride
Molecular Weight (g/mol): 414.58
Molecular Formula: C11H14BrClN3O5P
InChi Key: MZQQZBPMRPDKTB-UHFFFAOYNA-N
SMILES: Cl.CC(NCC1=C2NC(=O)C(=O)NC2=CC(Br)=C1)P(O)(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products