accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114162-64-0

(S)(S)O(R)(S)(S)ONHOOHHOHOClHOBrNH2

CAS RN 114162-64-0

IUPAC Name: (2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Synonyms: (2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular Weight (g/mol): 421.6
Molecular Formula: C14H12BrClNO7
InChi Key: DHJFFLKPAYHPHU-BYNIDDHOSA-M
SMILES: O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products