CAS RN 114214-69-6
CAS RN 114214-69-6
IUPAC Name:
tert-butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate
Synonyms:
tert-butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular Weight (g/mol):
201.27
Molecular Formula:
C10H19NO3
InChi Key:
HKIGXXRMJFUUKV-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)N1CCC(CO)C1
1
–
1
of
1
results
1
N-Boc-DL-beta-prolinol, 97+%, Thermo Scientific™
CAS: 114214-69-6 Molecular Formula: C10H19NO3 Molecular Weight (g/mol): 201.266 MDL Number: MFCD02179040 InChI Key: HKIGXXRMJFUUKV-UHFFFAOYSA-N Synonym: tert-butyl 3-hydroxymethyl pyrrolidine-1-carboxylate,1-boc-3-hydroxymethylpyrrolidine,1-boc-3-hydroxymethyl pyrrolidine,3-hydroxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester,3-hydroxymethyl-1-boc-pyrrolidine,1-boc-3-pyrrolidinemethanol,n-boc-dl-beta-prolinol,1-pyrrolidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester,tert-butyl 3-hydroxymethyl-1-pyrrolidinecarboxylate,3-hydroxymethylpyrrolidine-1-carboxylic acid tert-butyl ester PubChem CID: 5237175 IUPAC Name: tert-butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CCC(C1)CO