accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114382-20-6

NHNBr

CAS RN 114382-20-6

IUPAC Name: 5-(2-bromophenyl)-1H-pyrazole
Synonyms: 3-(2-bromophenyl)-2H-pyrazole
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: VDHTYVJAQNZWTD-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1=CC=NN1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products