man

Test

CAS RN 114821-24-8

NNI

CAS RN 114821-24-8

IUPAC Name: 2-iodopyridine-4-carbonitrile
Synonyms: 2-iodopyridine-4-carbonitrile
Molecular Weight (g/mol): 230.01
Molecular Formula: C6H3IN2
InChi Key: RAMAPMJJBMFFEI-UHFFFAOYSA-N
SMILES: IC1=NC=CC(=C1)C#N

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results