accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 114971-53-8

CH3ICH3OH

CAS RN 114971-53-8

IUPAC Name: 4-iodo-2,5-dimethylphenol
Synonyms: 4-iodo-2,5-dimethylphenol
Molecular Weight (g/mol): 248.06
Molecular Formula: C8H9IO
InChi Key: RFAXNKKOQUYDKE-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(C)C=C1I

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products