CAS RN 1150153-86-8
CAS RN 1150153-86-8
IUPAC Name:
1-{9-methyl-9-azabicyclo[3.3.1]nonan-3-yl}-2,3-dihydro-1H-indol-2-one
Synonyms:
1-{9-methyl-9-azabicyclo[3.3.1]nonan-3-yl}-3H-indol-2-one
Molecular Weight (g/mol):
270.38
Molecular Formula:
C17H22N2O
InChi Key:
GIDZCNCCCWFCIN-UHFFFAOYNA-N
SMILES:
CN1C2CCCC1CC(C2)N1C(=O)CC2=CC=CC=C12
1
–
1
of
1
results
1
Tocris Bioscience™ SR 16584

CAS: 1150153-86-8 Molecular Formula: C17H22N2O Molecular Weight (g/mol): 270.376