CAS RN 115436-72-1
CAS RN 115436-72-1
IUPAC Name:
sodium hydrogen [1-hydroxy-1-phosphono-2-(pyridin-3-yl)ethyl]phosphonate
Synonyms:
sodium hydrogen 1-hydroxy-1-phosphono-2-(pyridin-3-yl)ethylphosphonate
Molecular Weight (g/mol):
305.09
Molecular Formula:
C7H10NNaO7P2
InChi Key:
DRFDPXKCEWYIAW-UHFFFAOYNA-M
SMILES:
[Na+].OC(CC1=CN=CC=C1)(P(O)(O)=O)P(O)([O-])=O
1
–
3
of
3
results
1
Thermo Scientific Acros Risedronic acid sodium salt, Thermo Scientific Chemicals
CAS: 115436-72-1 Molecular Formula: C7H10NNaO7P2 Molecular Weight (g/mol): 305.09 MDL Number: MFCD00867080,MFCD01706268,MFCD00867080 InChI Key: DRFDPXKCEWYIAW-UHFFFAOYNA-M IUPAC Name: sodium hydrogen [1-hydroxy-1-phosphono-2-(pyridin-3-yl)ethyl]phosphonate SMILES: [Na+].OC(CC1=CN=CC=C1)(P(O)(O)=O)P(O)([O-])=O
2
Medchem Express Risedronate sodium, MedChemExpress
MedChemExpress Risedronate sodium is a pyridinyl biphosphonate which inhibits osteoclast-mediated bone resorption.
