accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 115913-58-1

CH3MgBr

CAS RN 115913-58-1

IUPAC Name: magnesium(2+) 4-methylnaphthalen-1-ide bromide
Synonyms: magnesium(2+) 4-methylnaphthalen-1-ide bromide
Molecular Weight (g/mol): 245.4
Molecular Formula: C11H9BrMg
InChi Key: JJEPJVHQVFSQDI-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC1=C2C=CC=CC2=[C-]C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products