accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 116057-81-9

H3CONCH3H3C

CAS RN 116057-81-9

IUPAC Name: N,N-dimethylacetamide
Synonyms: dimethylacetamide hallucinogen N,N'-dimethylacetamide
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: FXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILES: CN(C)C(C)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products