CAS RN 116247-03-1
CAS RN 116247-03-1
IUPAC Name:
2-(pyridin-3-yl)-4,5-dihydro-1,3-thiazole-4-carboxylic acid
Synonyms:
2-(pyridin-3-yl)-4,5-dihydro-1,3-thiazole-4-carboxylic acid
Molecular Weight (g/mol):
208.24
Molecular Formula:
C9H8N2O2S
InChi Key:
LJGAQGZEJDQDAU-UHFFFAOYNA-N
SMILES:
OC(=O)C1CSC(=N1)C1=CN=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Maybridge 2-Pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid, ≥97%, Thermo Scientific™
CAS: 116247-03-1 Molecular Formula: C9H8N2O2S Molecular Weight (g/mol): 208.24 MDL Number: MFCD02854711 InChI Key: LJGAQGZEJDQDAU-UHFFFAOYNA-N Synonym: 2-pyrid-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid,2-pyridin-3-yl-4,5-dihydro-1,3-thiazole-4-carboxylic acid,2-pyridin-3-yl-4,5-dihydro-thiazole-4-carboxylic acid,4-thiazolecarboxylicacid, 4,5-dihydro-2-3-pyridinyl,2-pyridin-3-yl-4,5-dihydrothiazole-4-carboxylic acid,3-4-carboxy-4,5-dihydro-1,3-thiazol-2-yl pyridine,2-3-pyridyl-1,3-thiazoline-4-carboxylic acid,acmc-20dw5p,4-carboxy-4,5-dihydro-2-pyridin-3-yl-1,3-thiazole PubChem CID: 3130734 SMILES: OC(=O)C1CSC(=N1)C1=CN=CC=C1