accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 116355-16-9

NNO

CAS RN 116355-16-9

IUPAC Name: imidazo[1,2-a]pyridine-6-carbaldehyde
Synonyms: imidazo[1,2-a]pyridine-6-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: VDWYLKPRUHCLAW-UHFFFAOYSA-N
SMILES: O=CC1=CN2C=CN=C2C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products
1x1 image pixel for data collection