accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 116632-39-4

CH3BrI

CAS RN 116632-39-4

IUPAC Name: 4-bromo-1-iodo-2-methylbenzene
Synonyms: 4-bromo-1-iodo-2-methylbenzene
Molecular Weight (g/mol): 296.93
Molecular Formula: C7H6BrI
InChi Key: GHTUADBHTFHMNI-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=C1I
Boiling Point 
  • (3)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products