accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 116821-45-5

H3COSiNH2OH3COCH3

CAS RN 116821-45-5

IUPAC Name: (11-aminoundecyl)triethoxysilane
Synonyms: (11-aminoundecyl)triethoxysilane
Molecular Weight (g/mol): 333.59
Molecular Formula: C17H39NO3Si
InChi Key: LPWZCJFZJCOBHO-UHFFFAOYSA-N
SMILES: CCO[Si](CCCCCCCCCCCN)(OCC)OCC

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products