accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 117414-74-1

HOO(R)NHNPOHOHO(E)

CAS RN 117414-74-1

IUPAC Name: (2R)-4-[(2E)-3-phosphonoprop-2-en-1-yl]piperazine-2-carboxylic acid
Synonyms: midafotel
Molecular Weight (g/mol): 250.19
Molecular Formula: C8H15N2O5P
InChi Key: VZXMZMJSGLFKQI-ABVWVHJUSA-N
SMILES: OC(=O)[C@H]1CN(C\C=C\P(O)(O)=O)CCN1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products