man

Test

CAS RN 117923-34-9

CAS RN 117923-34-9

IUPAC Name: 1-[2-(4-ethylphenyl)ethynyl]-4-hexylbenzene
Synonyms: 1-[2-(4-ethylphenyl)ethynyl]-4-hexylbenzene
Molecular Weight (g/mol): 290.45
Molecular Formula: C22H26
InChi Key: ZCEJWGXSGOFJTA-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)C#CC1=CC=C(CC)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)