man

Test

CAS RN 1184-78-7

CH3NCH3CH3O

CAS RN 1184-78-7

IUPAC Name: N,N-dimethylmethanamine oxide
Synonyms: trimethylamine-n-oxide
Molecular Weight (g/mol): 75.11
Molecular Formula: C3H9NO
InChi Key: UYPYRKYUKCHHIB-UHFFFAOYSA-N
SMILES: C[N+](C)(C)[O-]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results