accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1190-92-7

CH3NH3CNOO(E/Z)

CAS RN 1190-92-7

IUPAC Name: dimethyl(2-nitroethenyl)amine
Synonyms: dimethyl(2-nitroethenyl)amine
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: JKOVQYWMFZTKMX-UHFFFAOYSA-N
SMILES: CN(C)C=C[N+]([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products