man

Test

CAS RN 1190376-20-5

BOOH3CCH3H3CH3C

CAS RN 1190376-20-5

IUPAC Name: 4,4,5,5-tetramethyl-2-[4-(2-phenylethynyl)phenyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[4-(2-phenylethynyl)phenyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 304.2
Molecular Formula: C20H21BO2
InChi Key: AAIDERXZGJSORP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#CC1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results