CAS RN 119269-24-8
CAS RN 119269-24-8
IUPAC Name:
3-hexadecylthiophene
Synonyms:
3-hexadecylthiophene
Molecular Weight (g/mol):
308.57
Molecular Formula:
C20H36S
InChi Key:
FRVZSODZVJPMKO-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCC1=CSC=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 3-n-Hexadecylthiophene, 97%
CAS: 119269-24-8 Molecular Formula: C20H36S Molecular Weight (g/mol): 308.57 MDL Number: MFCD07368973 InChI Key: FRVZSODZVJPMKO-UHFFFAOYSA-N Synonym: 3-n-hexadecylthiophene,thiophene, 3-hexadecyl,3-hexadecyl-thiophene,ksc173e6j,acmc-1c275 PubChem CID: 2769638 IUPAC Name: 3-hexadecylthiophene SMILES: CCCCCCCCCCCCCCCCC1=CSC=C1