accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 119670-30-3

CH3CH3(S)HNOOONHO(S)HNONNCH3H3C

CAS RN 119670-30-3

IUPAC Name: benzyl N-[(1S)-1-{[(2S)-2-[(3-diazo-2-oxopropyl)amino]-3-methylbutanoyl]carbamoyl}-3-methylbutyl]carbamate
Synonyms: benzyl N-[(1S)-1-{[(2S)-2-[(3-diazo-2-oxopropyl)amino]-3-methylbutanoyl]carbamoyl}-3-methylbutyl]carbamate
Molecular Weight (g/mol): 445.52
Molecular Formula: C22H31N5O5
InChi Key: ZLDBPOCJCXPVND-OALUTQOASA-N
SMILES: CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)NC(=O)[C@@H](NCC(=O)C=[N+]=[N-])C(C)C
Product Type 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products