man

Test

CAS RN 12003-13-3

AlClClClClClClClTi

CAS RN 12003-13-3

IUPAC Name: titanium(4+) aluminium(3+) heptachloride
Synonyms: titanium(4+) aluminium(3+) heptachloride
Molecular Weight (g/mol): 323
Molecular Formula: AlCl7Ti
InChi Key: ZMKAVICCBWPNSR-UHFFFAOYSA-G
SMILES: [Al+3].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)