CAS RN 1202-34-2
CAS RN 1202-34-2
IUPAC Name:
N-(pyridin-2-yl)pyridin-2-amine
Synonyms:
N-2-pyridinyl-2-pyridinamine
Molecular Weight (g/mol):
171.2
Molecular Formula:
C10H9N3
InChi Key:
HMMPCBAWTWYFLR-UHFFFAOYSA-N
SMILES:
N(C1=CC=CC=N1)C1=CC=CC=N1