accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12031-83-3

LiLiOZrOO

CAS RN 12031-83-3

IUPAC Name: dilithium(1+) oxozirconiumbis(olate)
Synonyms: dilithium(1+) oxozirconiumbis(olate)
Molecular Weight (g/mol): 153.1
Molecular Formula: Li2O3Zr
InChi Key: PSKLXRHTVCSWPJ-UHFFFAOYSA-N
SMILES: [Li+].[Li+].[O-][Zr]([O-])=O
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products