man

Test

CAS RN 12034-77-4

SeNbSe

CAS RN 12034-77-4

IUPAC Name: diselanylideneniobium
Synonyms: diselanylideneniobium
Molecular Weight (g/mol): 250.85
Molecular Formula: NbSe2
InChi Key: CXRFFSKFQFGBOT-UHFFFAOYSA-N
SMILES: [Se]=[Nb]=[Se]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results