CAS RN 12047-11-9
CAS RN 12047-11-9
IUPAC Name:
dodecairon(3+) barium(2+) nonadecaoxidandiide
Synonyms:
dodecairon(3+) barium(2+) nonadecaoxidandiide
Molecular Weight (g/mol):
1111.45
Molecular Formula:
BaFe12O19
InChi Key:
HPYIMVBXZPJVBV-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[Ba++]
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Barium dodecairon nonadecaoxide
CAS: 12047-11-9 Molecular Formula: BaFe12O19 MDL Number: MFCD00075637 Synonym: Barium ferrite