CAS RN 121-02-8
CAS RN 121-02-8
IUPAC Name:
2-methyl-5-nitrobenzene-1-sulfonyl chloride
Synonyms:
2-methyl-5-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol):
235.64
Molecular Formula:
C7H6ClNO4S
InChi Key:
WPGVQDHXOUAJBW-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1S(Cl)(=O)=O)[N+]([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 2-Methyl-5-nitrobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
CAS: 121-02-8 Molecular Formula: C7H6ClNO4S Molecular Weight (g/mol): 235.64 MDL Number: MFCD00051695 InChI Key: WPGVQDHXOUAJBW-UHFFFAOYSA-N Synonym: 2-methyl-5-nitrobenzene-1-sulfonyl chloride,5-nitro-o-toluenesulfonyl chloride,2-methyl-5-nitrobenzenesulfonylchloride,benzenesulfonyl chloride, 2-methyl-5-nitro,4-nitrotoluen-2-sulfochlorid,o-toluenesulfonyl chloride, 5-nitro,4-nitrotoluen-2-sulfonylchlorid,4-nitrotoluene-2-sulphonyl chloride,4-nitrotoluen-2-sulfochlorid czech,2-methyl-5-nitrophenylsulfonyl chloride PubChem CID: 8457 IUPAC Name: 2-methyl-5-nitrobenzenesulfonyl chloride SMILES: CC1=CC=C(C=C1S(Cl)(=O)=O)[N+]([O-])=O