man

Test

CAS RN 121-45-9

CH3OPOH3COH3C

CAS RN 121-45-9

IUPAC Name: trimethyl phosphite
Synonyms: trimethyl phosphite
Molecular Weight (g/mol): 124.08
Molecular Formula: C3H9O3P
InChi Key: CYTQBVOFDCPGCX-UHFFFAOYSA-N
SMILES: COP(OC)OC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)