accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 121-75-5

H3COOSPSOCH3OCH3OOH3C

CAS RN 121-75-5

IUPAC Name: 1,4-diethyl 2-{[dimethoxy(sulfanylidene)-λ⁵-phosphanyl]sulfanyl}butanedioate
Synonyms: paladin phosphothion kop-thionkypfosmalacide kill-A-mite staeubol-puder kop-thion ortho malathion calmathion malathion (substance) 马拉硫磷
Molecular Weight (g/mol): 330.35
Molecular Formula: C10H19O6PS2
InChi Key: JXSJBGJIGXNWCI-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC
Molecular Weight (g/mol) 
  • (3)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
Concentration 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products