man

Test

CAS RN 121-81-3

NH2ONOONOO

CAS RN 121-81-3

IUPAC Name: 3,5-dinitrobenzamide
Synonyms: 3,5-dinitrobenzamide
Molecular Weight (g/mol): 211.13
Molecular Formula: C7H5N3O5
InChi Key: UUKWKUSGGZNXGA-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results