man

Test

CAS RN 121343-82-6

OONHOOHOHO

CAS RN 121343-82-6

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanedioic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanedioic acid
Molecular Weight (g/mol): 369.37
Molecular Formula: C20H19NO6
InChi Key: QEPWHIXHJNNGLU-UHFFFAOYNA-N
SMILES: OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results