accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12145-47-0

PdBrBr

CAS RN 12145-47-0

IUPAC Name: cycloocta-1,5-diene; dibromopalladium
Synonyms: 1,5-cyclooctadiene; palladium(II) bromide
Molecular Weight (g/mol): 374.41
Molecular Formula: C8H12Br2Pd
InChi Key: VDWGPWKGFNEZFD-UHFFFAOYSA-L
SMILES: Br[Pd]Br.C1CC=CCCC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products