CAS RN 1215034-77-7
CAS RN 1215034-77-7
IUPAC Name:
2,3-difluoro-4-(trifluoromethyl)pyridine
Synonyms:
2,3-difluoro-4-(trifluoromethyl)pyridine
Molecular Weight (g/mol):
183.08
Molecular Formula:
C6H2F5N
InChi Key:
ZOMVJIOHXSXCIP-UHFFFAOYSA-N
SMILES:
FC1=NC=CC(=C1F)C(F)(F)F
1
–
1
of
1
results
1
Thermo Scientific Acros 2,3-Difluoro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
CAS: 1215034-77-7 Molecular Formula: C6H2F5N Molecular Weight (g/mol): 183.07 InChI Key: ZOMVJIOHXSXCIP-UHFFFAOYSA-N Synonym: 2,3-difluoro-4-trifluoromethyl pyridine,alpha,alpha,alpha,2,3-pentafluoro-4-picoline PubChem CID: 13491638 IUPAC Name: 2,3-difluoro-4-(trifluoromethyl)pyridine SMILES: C1=CN=C(C(=C1C(F)(F)F)F)F