CAS RN 1215333-48-4
CAS RN 1215333-48-4
IUPAC Name:
2-{4-[4-(4-carbamimidoylphenyl)piperazin-1-yl]piperidin-1-yl}acetic acid trihydrochloride
Synonyms:
{4-[4-(4-carbamimidoylphenyl)piperazin-1-yl]piperidin-1-yl}acetic acid trihydrochloride
Molecular Weight (g/mol):
454.82
Molecular Formula:
C18H30Cl3N5O2
InChi Key:
YKRNPHOBDOUQTG-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.NC(=N)C1=CC=C(C=C1)N1CCN(CC1)C1CCN(CC(O)=O)CC1
1
–
1
of
1
results
1
GR 144053 trihydrochloride, Tocris Bioscience™
CAS: 1215333-48-4 Molecular Formula: C18H30Cl3N5O2 Molecular Weight (g/mol): 454.821 InChI Key: YKRNPHOBDOUQTG-UHFFFAOYSA-N Synonym: gr 144053 trihydrochloride,4-4-4-aminoiminomethyl phenyl-1-piperazinyl-1-piperidineacetic acid trihydrochloride,2-4-4-4-carbamimidoylphenyl piperazin-1-yl piperidin-1-yl acetic acid trihydrochloride PubChem CID: 16759148 IUPAC Name: 2-[4-[4-(4-carbamimidoylphenyl)piperazin-1-yl]piperidin-1-yl]acetic acid;trihydrochloride SMILES: C1CN(CCC1N2CCN(CC2)C3=CC=C(C=C3)C(=N)N)CC(=O)O.Cl.Cl.Cl