CAS RN 1215654-26-4
CAS RN 1215654-26-4
IUPAC Name:
5-methyl-3-(3-{4-[2-(2,2,2-trifluoroethoxy)phenyl]piperazin-1-yl}propyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride
Synonyms:
5-methyl-3-(3-{4-[2-(2,2,2-trifluoroethoxy)phenyl]piperazin-1-yl}propyl)-1H-pyrimidine-2,4-dione hydrochloride
Molecular Weight (g/mol):
462.9
Molecular Formula:
C20H26ClF3N4O3
InChi Key:
CWVABCXVOAVUJL-UHFFFAOYSA-N
SMILES:
Cl.CC1=CNC(=O)N(CCCN2CCN(CC2)C2=CC=CC=C2OCC(F)(F)F)C1=O
1
–
1
of
1
results
1
RS 100329 hydrochloride, Tocris Bioscience™
CAS: 1215654-26-4 Molecular Formula: C20H26ClF3N4O3 Molecular Weight (g/mol): 462.898 InChI Key: CWVABCXVOAVUJL-UHFFFAOYSA-N Synonym: rs 100329 hydrochloride,5-methyl-3-3-3-4-2-2,2,2-trifluoroethoxy phenyl-1-piperazinyl propyl-2,4-1h,3h-pyrimidinedione hydrochloride,5-methyl-3-3-3-4-2-2,2,2,-trifluroethoxy phenyl-1-piperazinyl propyl-2,4-1h,3h-pyrimidinedione hydrochloride PubChem CID: 11340200 IUPAC Name: 5-methyl-3-[3-[4-[2-(2,2,2-trifluoroethoxy)phenyl]piperazin-1-yl]propyl]-1H-pyrimidine-2,4-dione;hydrochloride SMILES: CC1=CNC(=O)N(C1=O)CCCN2CCN(CC2)C3=CC=CC=C3OCC(F)(F)F.Cl