CAS RN 1215766-76-9
CAS RN 1215766-76-9
IUPAC Name:
5-chloro-N4-(cyclopropylmethyl)-2-methyl-N4-propyl-N6-(2,4,6-trichlorophenyl)pyrimidine-4,6-diamine hydrochloride
Synonyms:
5-chloro-N4-(cyclopropylmethyl)-2-methyl-N4-propyl-N6-(2,4,6-trichlorophenyl)pyrimidine-4,6-diamine hydrochloride
Molecular Weight (g/mol):
470.64
Molecular Formula:
C18H21Cl5N4
InChi Key:
CPMGENCTAWBLNW-UHFFFAOYSA-N
SMILES:
Cl.CCCN(CC1CC1)C1=NC(C)=NC(NC2=C(Cl)C=C(Cl)C=C2Cl)=C1Cl
1
–
1
of
1
results
1
NBI 27914 hydrochloride, Tocris Bioscience™
CAS: 1215766-76-9 Molecular Formula: C18H21Cl5N4 Molecular Weight (g/mol): 470.644 InChI Key: CPMGENCTAWBLNW-UHFFFAOYSA-N Synonym: nbi 27914 hydrochloride,5-chloro-n-cyclopropylmethyl-2-methyl-n-propyl-n'-2,4,6-trichlorophenyl-4,6-pyrimidinediamine hydrochloride,5-chloro-4-n-cyclopropyl methyl-n-propylamino-2-methyl-6-2,4,6-trichlorophenyl aminopyridine PubChem CID: 45073446 IUPAC Name: 5-chloro-4-N-(cyclopropylmethyl)-2-methyl-4-N-propyl-6-N-(2,4,6-trichlorophenyl)pyrimidine-4,6-diamine;hydrochloride SMILES: CCCN(CC1CC1)C2=NC(=NC(=C2Cl)NC3=C(C=C(C=C3Cl)Cl)Cl)C.Cl