CAS RN 1217457-86-7
CAS RN 1217457-86-7
IUPAC Name:
methyl 2-({4-[(2-amino-3-sulfanylpropyl)amino]-2-(naphthalen-1-yl)phenyl}formamido)-4-methylpentanoate; trifluoroacetic acid
Synonyms:
methyl 2-({4-[(2-amino-3-sulfanylpropyl)amino]-2-(naphthalen-1-yl)phenyl}formamido)-4-methylpentanoate; trifluoroacetic acid
Molecular Weight (g/mol):
593.66
Molecular Formula:
C29H34F3N3O5S
InChi Key:
WALKWJPZELDSKT-UHFFFAOYNA-N
SMILES:
OC(=O)C(F)(F)F.COC(=O)C(CC(C)C)NC(=O)C1=C(C=C(NCC(N)CS)C=C1)C1=C2C=CC=CC2=CC=C1