CAS RN 1217674-10-6
CAS RN 1217674-10-6
IUPAC Name:
N,3-dimethyl-N-[4-(4-methylpiperidin-1-yl)butan-2-yl]benzene-1-sulfonamide hydrochloride
Synonyms:
N,3-dimethyl-N-[4-(4-methylpiperidin-1-yl)butan-2-yl]benzenesulfonamide hydrochloride
Molecular Weight (g/mol):
374.97
Molecular Formula:
C18H31ClN2O2S
InChi Key:
UIZKHTBWJSUGOV-UHFFFAOYNA-N
SMILES:
Cl.CC(CCN1CCC(C)CC1)N(C)S(=O)(=O)C1=CC=CC(C)=C1
1
–
1
of
1
results
1
SB 258719 hydrochloride, Tocris Bioscience™
CAS: 1217674-10-6 Molecular Formula: C18H31ClN2O2S Molecular Weight (g/mol): 374.968 InChI Key: UIZKHTBWJSUGOV-UNTBIKODSA-N Synonym: sb 258719 hydrochloride,3-methyl-n-1r-1-methyl-3-4-methyl-1-piperidinyl propyl-n-methylbenzenesulfonamide hydrochloride,n,3-dimethyl-n-2r-4-4-methylpiperidin-1-yl butan-2-yl benzenesulfonamide hydrochloride PubChem CID: 56972181 IUPAC Name: N,3-dimethyl-N-[(2R)-4-(4-methylpiperidin-1-yl)butan-2-yl]benzenesulfonamide;hydrochloride SMILES: CC1CCN(CC1)CCC(C)N(C)S(=O)(=O)C2=CC=CC(=C2)C.Cl