accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12323-19-2

SbSbBiBi

CAS RN 12323-19-2

IUPAC Name: 1,2-distiba-3,4-dibismatricyclo[1.1.0.0²,⁴]butane
Synonyms: 1,2-distiba-3,4-dibismatricyclo[1.1.0.0²,⁴]butane
Molecular Weight (g/mol): 661.48
Molecular Formula: Bi2Sb2
InChi Key: AEMQIQQWIVNHAU-UHFFFAOYSA-N
SMILES: [Sb]12[Sb]3[Bi]1[Bi]23

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products