CAS RN 12333-11-8
CAS RN 12333-11-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ammonium tungsten oxide hydrate, Thermo Scientific Chemicals
CAS: 12333-11-8 Molecular Formula: (NH4)6W12O39·xH2O MDL Number: MFCD00042673 Synonym: Ammonium metatungstate hydrate