accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1235-82-1

HClOHN

CAS RN 1235-82-1

IUPAC Name: hydrogen 1-{bicyclo[2.2.1]hept-5-en-2-yl}-1-phenyl-3-(piperidin-1-yl)propan-1-ol chloride
Synonyms: hydrogen biperiden chloride
Molecular Weight (g/mol): 347.93
Molecular Formula: C21H30ClNO
InChi Key: RDNLAULGBSQZMP-UHFFFAOYNA-N
SMILES: [H+].[Cl-].OC(CCN1CCCCC1)(C1CC2CC1C=C2)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products