man

Test

CAS RN 123732-04-7

H3COBr

CAS RN 123732-04-7

IUPAC Name: 1-bromo-4-(heptyloxy)benzene
Synonyms: 1-bromo-4-(heptyloxy)benzene
Molecular Weight (g/mol): 271.2
Molecular Formula: C13H19BrO
InChi Key: DSCVEUSJTAFLBF-UHFFFAOYSA-N
SMILES: CCCCCCCOC1=CC=C(Br)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results