man

Test

CAS RN 1240257-01-5

CH3OSFFFFF

CAS RN 1240257-01-5

IUPAC Name: 3-methyl-5-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Synonyms: 3-methyl-5-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Molecular Weight (g/mol): 246.2
Molecular Formula: C8H7F5OS
InChi Key: MBCKIHKPQIVSBW-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C=O)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results