man

Test

CAS RN 1240257-02-6

SFFFFFOF

CAS RN 1240257-02-6

IUPAC Name: 2-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Synonyms: 2-fluoro-5-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Molecular Weight (g/mol): 250.16
Molecular Formula: C7H4F6OS
InChi Key: LWVYGZGWRNJVBG-UHFFFAOYSA-N
SMILES: FC1=C(C=O)C=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results