man

Test

CAS RN 1240257-88-8

SFFFFFClO

CAS RN 1240257-88-8

IUPAC Name: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Synonyms: 3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Molecular Weight (g/mol): 266.61
Molecular Formula: C7H4ClF5OS
InChi Key: AHEFJQKJBDJLSO-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC(Cl)=CC(C=O)=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results