man

Test

CAS RN 1240257-95-7

SFFFFFBrNH2

CAS RN 1240257-95-7

IUPAC Name: 3-bromo-5-(pentafluoro-λ⁶-sulfanyl)aniline
Synonyms: 3-bromo-5-(pentafluoro-λ⁶-sulfanyl)aniline
Molecular Weight (g/mol): 298.07
Molecular Formula: C6H5BrF5NS
InChi Key: HCROKBUNMALNHZ-UHFFFAOYSA-N
SMILES: NC1=CC(=CC(Br)=C1)S(F)(F)(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results