CAS RN 124655-17-0
CAS RN 124655-17-0
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]-3,3-dimethylbutanoic acid
Molecular Weight (g/mol):
231.29
Molecular Formula:
C11H21NO4
InChi Key:
LRFZIPCTFBPFLX-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(C(O)=O)C(C)(C)C
1
–
1
of
1
results
1
N-Boc-D-tert-leucine, 95%, Thermo Scientific Chemicals
CAS: 124655-17-0 Molecular Formula: C11H21NO4 Molecular Weight (g/mol): 231.29 MDL Number: MFCD00065575 InChI Key: LRFZIPCTFBPFLX-JLDDOWRYNA-N Synonym: boc-d-tert-leucine,n-boc-d-tert-leucine,boc-d-tle-oh,r-n-tert-butoxycarbonyl-tert-leucine,r-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid,boc-d-alpha-t-butylglycine,boc-tbu-d-gly-oh,boc-d-t-leu,d-valine, n-1,1-dimethylethoxy carbonyl-3-methyl,n-tert-butoxycarbonyl-d-tert-leucine PubChem CID: 7005057 SMILES: CC(C)(C)OC(=O)N[C@@H](C(O)=O)C(C)(C)C